Product Details

SNP ID
rs7259253
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:13126688 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGACTGCTGGCTACCCACCTTGGCA[A/G]CTGTCAAAGGAGGTCGTCTGTCTGC
Phenotype
MIM: 610672 MIM: 611669
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NACC1 PubMed Links

Gene Details

Gene
NACC1
Gene Name
nucleus accumbens associated 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052876.3 Intron NP_443108.1
XM_005259721.3 Intron XP_005259778.1
Gene
TRMT1
Gene Name
tRNA methyltransferase 1
There are no transcripts associated with this gene.

View Full Product Details