Product Details
- SNP ID
-
rs9891306
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:34576507 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTGAGACAGAGTCGGATTCCAGGAT[A/G]ATCCCAGGAACATGTCTTCATGATG
- Phenotype
-
MIM: 616178
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
C17orf102
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs79703349] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C17orf102
- Gene Name
- chromosome 17 open reading frame 102
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_207454.2 |
1613 |
UTR 3 |
|
|
NP_997337.2 |
- Gene
- TMEM132E
- Gene Name
- transmembrane protein 132E
There are no transcripts associated with this gene.
View Full Product Details