Product Details

SNP ID
rs11883056
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:3770427 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCGCTGCCAGCAGGACGGGTGGC[A/G]ATGGGGTGGGAGACGGGATGGGGGC
Phenotype
MIM: 604262 MIM: 600038 MIM: 611858 MIM: 610362
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
APBA3 PubMed Links

Gene Details

Gene
APBA3
Gene Name
amyloid beta precursor protein binding family A member 3
There are no transcripts associated with this gene.

Gene
MATK
Gene Name
megakaryocyte-associated tyrosine kinase
There are no transcripts associated with this gene.

Gene
MRPL54
Gene Name
mitochondrial ribosomal protein L54
There are no transcripts associated with this gene.

Gene
RAX2
Gene Name
retina and anterior neural fold homeobox 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319074.1 817 UTR 3 NP_001306003.1
NM_032753.3 817 UTR 3 NP_116142.1

View Full Product Details