Product Details

SNP ID
rs2240774
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:76714490 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGTACCCACTCTTCCTAGTCTACC[C/T]TGGCCACCCGCCACCACTCCACTCC
Phenotype
MIM: 604914
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
JMJD6 PubMed Links

Gene Details

Gene
JMJD6
Gene Name
arginine demethylase and lysine hydroxylase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001081461.1 3765 UTR 3 NP_001074930.1
NM_015167.2 3765 Intron NP_055982.2
Gene
MXRA7
Gene Name
matrix remodeling associated 7
There are no transcripts associated with this gene.

View Full Product Details