Product Details

SNP ID
rs4729132
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.7:94399612 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTGTATAACACTTTTGCCAATAT[A/G]TGAATACCTATATCTTATATCTATT
Phenotype
MIM: 120160
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
COL1A2 PubMed Links

Gene Details

Gene
COL1A2
Gene Name
collagen type I alpha 2 chain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000089.3 Intron NP_000080.2

View Full Product Details