Product Details

SNP ID
rs1003645
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.17:36013244 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CGTGTGTCCAGCTTCAGCATTCTCA[C/T]GCAAACCTGAACTTGCTTATCACTG
Phenotype
MIM: 602494
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCL23 PubMed Links

Gene Details

Gene
CCL23
Gene Name
C-C motif chemokine ligand 23
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005064.5 438 Missense Mutation ATG,GTG M123V NP_005055.3
NM_145898.3 438 Missense Mutation ATG,GTG M106V NP_665905.2

View Full Product Details