Product Details

SNP ID
rs1006767
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:35488960 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAGTCCCCGAAAGAACGCAACCC[A/G]TTCCCCACCATGGTCTCCGTTCCAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DMKN PubMed Links

Gene Details

Gene
DMKN
Gene Name
dermokine
There are no transcripts associated with this gene.

Gene
KRTDAP
Gene Name
keratinocyte differentiation associated protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001244847.1 Intron NP_001231776.1
NM_207392.2 Intron NP_997275.1

View Full Product Details