Product Details

SNP ID
rs4262
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.7:93922116 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AGCGGCTCCGCTGCCAGAGCTAGCC[C/T]GAGCCCGGTTCTGGGGCGAAAATGC
Phenotype
MIM: 604390
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GNG11 PubMed Links

Gene Details

Gene
GNG11
Gene Name
G protein subunit gamma 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004126.3 413 UTR 5 NP_004117.1

View Full Product Details