Product Details

SNP ID
rs2529541
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:30218761 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCTCCTGCTGCTGGCATTCCCCAG[A/G]AGCCTCAGAGAGCCCCAACCACTGC
Phenotype
MIM: 300098
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MAGEB2 PubMed Links

Gene Details

Gene
MAGEB2
Gene Name
MAGE family member B2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002364.4 324 Missense Mutation AAG,GAG K61E NP_002355.2
XM_011545512.1 324 Missense Mutation AAG,GAG K61E XP_011543814.1

View Full Product Details