Product Details

SNP ID
rs1296810
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:17574304 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATTTAAAAATTCACTTTTCATGT[C/T]GTGAAAATGCCAATCAAAAAGAAAA
Phenotype
MIM: 609303
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC101929372 PubMed Links

Gene Details

Gene
LOC101929372
Gene Name
uncharacterized LOC101929372
There are no transcripts associated with this gene.

Gene
SLC25A18
Gene Name
solute carrier family 25 member 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303484.1 Intron NP_001290413.1
NM_031481.2 Intron NP_113669.1
XM_011546149.2 Intron XP_011544451.1
XM_011546150.2 Intron XP_011544452.1
XM_011546151.2 Intron XP_011544453.1
XM_011546152.1 Intron XP_011544454.1
XM_011546153.2 Intron XP_011544455.1
XM_011546154.2 Intron XP_011544456.1
XM_017028968.1 Intron XP_016884457.1
XM_017028969.1 Intron XP_016884458.1
XM_017028970.1 Intron XP_016884459.1
XM_017028971.1 Intron XP_016884460.1
XM_017028972.1 Intron XP_016884461.1
XM_017028973.1 Intron XP_016884462.1
XM_017028974.1 Intron XP_016884463.1
XM_017028975.1 Intron XP_016884464.1

View Full Product Details