Product Details
- SNP ID
-
rs12539833
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:141664051 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GCCTCTGTAAGGAGCCCGGCTCTAA[A/G]TGCCCTTGGCAGCAATAAACGGCAT
- Phenotype
-
MIM: 610345
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
AGK
PubMed Links
Gene Details
- Gene
- AGK
- Gene Name
- acylglycerol kinase
There are no transcripts associated with this gene.
- Gene
- KIAA1147
- Gene Name
- KIAA1147
View Full Product Details