Product Details
- SNP ID
-
rs9655950
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:87404245 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- TAAAAGCAAAAATAATTAAGCTCTA[C/T]GTCAGTGTAACCCAGTGGAACCCCG
- Phenotype
-
MIM: 171060
MIM: 606090
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ABCB4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs45536638] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ABCB4
- Gene Name
- ATP binding cassette subfamily B member 4
- Gene
- CROT
- Gene Name
- carnitine O-octanoyltransferase
There are no transcripts associated with this gene.
View Full Product Details