Product Details

SNP ID
rs13232120
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:73568980 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAAAAATTCTTTTGGATTTTATTCC[A/T]AGTGTGATGGGAAGCCTTGGGGGGT
Phenotype
MIM: 605842
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
TBL2 PubMed Links

Gene Details

Gene
TBL2
Gene Name
transducin (beta)-like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012453.2 3005 UTR 3 NP_036585.1
XM_006715923.3 3005 Intron XP_006715986.1
XM_011516027.2 3005 Intron XP_011514329.1

View Full Product Details