Product Details

SNP ID
rs853907
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.10:118312034 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTTTAACTCAACCTAAAACAGTCC[C/T]TGTTTCTCCAGCCCTCCAGCTTTCA
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
FAM204A PubMed Links

Gene Details

Gene
FAM204A
Gene Name
family with sequence similarity 204 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134672.1 Intron NP_001128144.1
NM_022063.2 Intron NP_071346.1
XM_005270024.1 Intron XP_005270081.1

View Full Product Details