Product Details

SNP ID
rs34927155
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.16:10946622 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
AGGAGGGGAGGGGAGTTATCCAGAG[A/G]AGGTTGGGCCTCCTGCCGGCATCCA
Phenotype
MIM: 600005 MIM: 611303
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CIITA PubMed Links

Gene Details

Gene
CIITA
Gene Name
class II, major histocompatibility complex, transactivator
There are no transcripts associated with this gene.

Gene
CLEC16A
Gene Name
C-type lectin domain family 16 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243403.1 Intron NP_001230332.1
NM_015226.2 Intron NP_056041.1
XM_005255210.1 Intron XP_005255267.1
XM_005255211.1 Intron XP_005255268.1
XM_005255213.1 Intron XP_005255270.1
XM_005255214.1 Intron XP_005255271.1
XM_005255215.3 Intron XP_005255272.1
XM_005255216.1 Intron XP_005255273.1
XM_006720870.3 Intron XP_006720933.1
XM_011522434.1 Intron XP_011520736.1
XM_011522435.1 Intron XP_011520737.1
XM_011522436.2 Intron XP_011520738.1
XM_011522437.2 Intron XP_011520739.1
XM_011522438.2 Intron XP_011520740.1
XM_011522439.2 Intron XP_011520741.1
XM_011522440.2 Intron XP_011520742.1
XM_017023089.1 Intron XP_016878578.1
XM_017023090.1 Intron XP_016878579.1
Gene
DEXI
Gene Name
Dexi homolog
There are no transcripts associated with this gene.

View Full Product Details