Product Details

SNP ID
rs12558624
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:114595563 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTTCCAGTACTTTCCCCTCTCTTC[A/G]TGTCATGGATATTTAGTTCCAGAGA
Phenotype
MIM: 312861
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HTR2C PubMed Links

Gene Details

Gene
HTR2C
Gene Name
5-hydroxytryptamine receptor 2C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000868.3 Intron NP_000859.1
NM_001256760.2 Intron NP_001243689.1
NM_001256761.2 Intron NP_001243690.1

View Full Product Details