Product Details
- SNP ID
-
rs10910031
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
32
- Location
-
Chr.1:2231568 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAGACAGGCCAGACAGGCTCTGGTC[G/A]TCTCAGCGGATGCCTCCGAGCCGCC
- Phenotype
-
MIM: 164780
- Polymorphism
- G/A, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
SKI
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs138776564] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SKI
- Gene Name
- SKI proto-oncogene
View Full Product Details