Product Details

SNP ID
rs3932173
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:179027886 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGCCCTGACTTTCTTCCTGGGACCA[C/T]CTCCTGGTTGTCCTCTAGGAGCCCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF879 PubMed Links

Gene Details

Gene
ZNF879
Gene Name
zinc finger protein 879
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136116.1 Intron NP_001129588.1
XM_005265908.3 Intron XP_005265965.1
XM_011534550.2 Intron XP_011532852.1
XM_011534551.2 Intron XP_011532853.1

View Full Product Details