Product Details

SNP ID
rs11788425
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:34333857 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCTTTACAGGCCAGCTATGCCCTT[A/G]TGCAAGTATTTTAACCTTTTGAAAT
Phenotype
MIM: 613747 MIM: 602852
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KIF24 PubMed Links

Gene Details

Gene
KIF24
Gene Name
kinesin family member 24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_194313.2 Intron NP_919289.2
XM_005251451.2 Intron XP_005251508.1
XM_006716764.2 Intron XP_006716827.1
XM_011517861.1 Intron XP_011516163.1
XM_011517863.2 Intron XP_011516165.1
XM_017014697.1 Intron XP_016870186.1
XM_017014698.1 Intron XP_016870187.1
Gene
NUDT2
Gene Name
nudix hydrolase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001161.4 Intron NP_001152.1
NM_001244390.1 Intron NP_001231319.1
NM_147172.2 Intron NP_671701.1
NM_147173.2 Intron NP_671702.1

View Full Product Details