Product Details

SNP ID
rs12006123
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:32456019 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TACTTGTTTCCACGTCCAGGAAACC[A/G]CAAACTAAGCTCTTTGAAGCATCAG
Phenotype
MIM: 100880 MIM: 609631
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACO1 PubMed Links

Gene Details

Gene
ACO1
Gene Name
aconitase 1
There are no transcripts associated with this gene.

Gene
DDX58
Gene Name
DEXD/H-box helicase 58
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014314.3 4039 UTR 3 NP_055129.2
Gene
LOC101060445
Gene Name
uncharacterized LOC101060445
There are no transcripts associated with this gene.

View Full Product Details