Product Details

SNP ID
rs4906191
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:102323151 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGAGTCCAAAGAGGGCCAGTTGGT[C/G]GTCCCAAATAAGTGATTCTCAACCG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZNF839 PubMed Links

Gene Details

Gene
ZNF839
Gene Name
zinc finger protein 839
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001267827.1 Intron NP_001254756.1
NM_001267828.1 Intron NP_001254757.1
NM_018335.4 Intron NP_060805.3
XM_006720203.3 Intron XP_006720266.2
XM_011536945.1 Intron XP_011535247.1
XM_011536946.2 Intron XP_011535248.1
XM_011536948.2 Intron XP_011535250.1
XM_011536949.2 Intron XP_011535251.1
XM_011536950.2 Intron XP_011535252.1
XM_017021447.1 Intron XP_016876936.1
XM_017021448.1 Intron XP_016876937.1
XM_017021449.1 Intron XP_016876938.1
XM_017021450.1 Intron XP_016876939.1
XM_017021451.1 Intron XP_016876940.1

View Full Product Details