Product Details

SNP ID
rs11631770
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:41444861 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCCATAGAGTTTAGGCTGGAATAC[C/T]GCGCTTTAGTACTTGAACTTAGTGA
Phenotype
MIM: 611633
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RTF1 PubMed Links

Gene Details

Gene
RTF1
Gene Name
RTF1 homolog, Paf1/RNA polymerase II complex component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015138.4 Intron NP_055953.3

View Full Product Details