Product Details

SNP ID
rs1877954
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.2:85427176 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CCTGTTCTCAGTAGCTGAGACCTGA[A/G]AAATTTGGAGAATCACTTTGTGCTA
Phenotype
MIM: 153615
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CAPG PubMed Links

Gene Details

Gene
CAPG
Gene Name
capping actin protein, gelsolin like
There are no transcripts associated with this gene.

Gene
SH2D6
Gene Name
SH2 domain containing 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_201594.2 1058 Intron NP_963888.2
XM_011532783.1 1058 Intron XP_011531085.1
XM_011532796.2 1058 Intron XP_011531098.1
XM_017003834.1 1058 UTR 5 XP_016859323.1
XM_017003835.1 1058 UTR 5 XP_016859324.1
XM_017003836.1 1058 UTR 5 XP_016859325.1
XM_017003837.1 1058 UTR 5 XP_016859326.1
XM_017003838.1 1058 UTR 5 XP_016859327.1
XM_017003839.1 1058 Intron XP_016859328.1
XM_017003840.1 1058 UTR 5 XP_016859329.1
XM_017003841.1 1058 UTR 5 XP_016859330.1
XM_017003842.1 1058 UTR 5 XP_016859331.1
XM_017003843.1 1058 UTR 5 XP_016859332.1
XM_017003844.1 1058 UTR 5 XP_016859333.1
XM_017003845.1 1058 UTR 5 XP_016859334.1
XM_017003846.1 1058 UTR 5 XP_016859335.1
XM_017003847.1 1058 UTR 5 XP_016859336.1
XM_017003848.1 1058 UTR 5 XP_016859337.1
XM_017003849.1 1058 UTR 5 XP_016859338.1
XM_017003850.1 1058 UTR 5 XP_016859339.1
XM_017003851.1 1058 UTR 5 XP_016859340.1
XM_017003852.1 1058 Intron XP_016859341.1
XM_017003853.1 1058 UTR 5 XP_016859342.1
XM_017003854.1 1058 UTR 5 XP_016859343.1
XM_017003855.1 1058 UTR 5 XP_016859344.1
XM_017003856.1 1058 UTR 5 XP_016859345.1
XM_017003857.1 1058 UTR 5 XP_016859346.1
XM_017003858.1 1058 Intron XP_016859347.1
XM_017003859.1 1058 Intron XP_016859348.1
XM_017003860.1 1058 Intron XP_016859349.1

View Full Product Details