Product Details

SNP ID
rs11678613
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:85421429 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATGGACAGGAAATGGGCAAGACAC[A/G]GGCCTTTCAGAGAAGAGAGACCAAA
Phenotype
MIM: 153615
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CAPG PubMed Links

Gene Details

Gene
CAPG
Gene Name
capping actin protein, gelsolin like
There are no transcripts associated with this gene.

Gene
SH2D6
Gene Name
SH2 domain containing 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_201594.2 Intron NP_963888.2
XM_011532783.1 Intron XP_011531085.1
XM_011532796.2 Intron XP_011531098.1
XM_017003834.1 Intron XP_016859323.1
XM_017003835.1 Intron XP_016859324.1
XM_017003836.1 Intron XP_016859325.1
XM_017003837.1 Intron XP_016859326.1
XM_017003838.1 Intron XP_016859327.1
XM_017003839.1 Intron XP_016859328.1
XM_017003840.1 Intron XP_016859329.1
XM_017003841.1 Intron XP_016859330.1
XM_017003842.1 Intron XP_016859331.1
XM_017003843.1 Intron XP_016859332.1
XM_017003844.1 Intron XP_016859333.1
XM_017003845.1 Intron XP_016859334.1
XM_017003846.1 Intron XP_016859335.1
XM_017003847.1 Intron XP_016859336.1
XM_017003848.1 Intron XP_016859337.1
XM_017003849.1 Intron XP_016859338.1
XM_017003850.1 Intron XP_016859339.1
XM_017003851.1 Intron XP_016859340.1
XM_017003852.1 Intron XP_016859341.1
XM_017003853.1 Intron XP_016859342.1
XM_017003854.1 Intron XP_016859343.1
XM_017003855.1 Intron XP_016859344.1
XM_017003856.1 Intron XP_016859345.1
XM_017003857.1 Intron XP_016859346.1
XM_017003858.1 Intron XP_016859347.1
XM_017003859.1 Intron XP_016859348.1
XM_017003860.1 Intron XP_016859349.1

View Full Product Details