Product Details

SNP ID
rs1544713
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:185741198 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTACTCTATTGCCATTTTCTCTGTC[A/G]ACCATTAATTCACTTGTCTACTACT
Phenotype
MIM: 615796
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FSIP2 PubMed Links

Gene Details

Gene
FSIP2
Gene Name
fibrous sheath interacting protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173651.3 Intron NP_775922.3
Gene
LOC101927196
Gene Name
uncharacterized LOC101927196
There are no transcripts associated with this gene.

View Full Product Details