Product Details

SNP ID
rs34311140
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:106584042 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGCTGTGACCTTGCTGGAGGGAGG[A/G]CAGGACGCAGACTGGCCCGCTAAAC
Phenotype
MIM: 603958
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC100287944 PubMed Links

Gene Details

Gene
LOC100287944
Gene Name
uncharacterized LOC100287944
There are no transcripts associated with this gene.

Gene
RFX4
Gene Name
regulatory factor X4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001206691.1 Intron NP_001193620.1
NM_032491.5 Intron NP_115880.2
NM_213594.2 Intron NP_998759.1

View Full Product Details