Product Details

SNP ID
rs2430821
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:139605825 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATGGGCTGGATGTCTACTTGGAC[A/T]TGTAAACATTAGTTACTAGTTTTTT
Phenotype
MIM: 605874
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
KCNK9 PubMed Links

Gene Details

Gene
KCNK9
Gene Name
potassium two pore domain channel subfamily K member 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282534.1 Intron NP_001269463.1
XM_011517101.2 Intron XP_011515403.1
XM_011517102.2 Intron XP_011515404.1
XM_011517103.2 Intron XP_011515405.1
XM_017013530.1 Intron XP_016869019.1
XM_017013531.1 Intron XP_016869020.1

View Full Product Details