Product Details

SNP ID
rs2824303
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:17447035 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGCCAAACAAAGGGCCCTCCAGTTA[C/G]CAGAAATTGCCCTCCTCAGAAGAAA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LINC01549 PubMed Links

Gene Details

Gene
LINC01549
Gene Name
long intergenic non-protein coding RNA 1549
There are no transcripts associated with this gene.

View Full Product Details