Product Details

SNP ID
rs564253
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:116103551 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCACCCAGAGGAGTTAGAGTAAAGG[C/T]CAGAACCTCGAGGTGAGCAACAGCA
Phenotype
MIM: 606573
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FRK PubMed Links

Gene Details

Gene
FRK
Gene Name
fyn related Src family tyrosine kinase
There are no transcripts associated with this gene.

Gene
NT5DC1
Gene Name
5'-nucleotidase domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152729.2 Intron NP_689942.2
XM_006715378.3 Intron XP_006715441.1

View Full Product Details