Product Details

SNP ID
rs8111699
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:1209715 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGCCTGACTGTGAGAGTGAGCCCC[C/G]TGCTTTCCTGAACAGTCAGTGGAGG
Phenotype
MIM: 602216
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
STK11 PubMed Links

Gene Details

Gene
STK11
Gene Name
serine/threonine kinase 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000455.4 Intron NP_000446.1
XM_005259617.2 Intron XP_005259674.1
XM_005259618.3 Intron XP_005259675.1
XM_011528209.1 Intron XP_011526511.1

View Full Product Details