Product Details

SNP ID
rs13160737
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:32356870 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGAGTACTAACAAATGAAGCTCCCC[A/C]AAATTTATACTGATTAATATATCCA
Phenotype
MIM: 615635
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ZFR PubMed Links

Gene Details

Gene
ZFR
Gene Name
zinc finger RNA binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016107.3 Intron NP_057191.2

View Full Product Details