Product Details

SNP ID
rs287103
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.19:33801816 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CCTGGGAAATTCACAAGGGGAGGGA[A/G]GAGGAGCTGTGGTGAGCTGCAGTGT
Phenotype
MIM: 615240
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KCTD15 PubMed Links

Gene Details

Gene
KCTD15
Gene Name
potassium channel tetramerization domain containing 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001129994.1 Intron NP_001123466.1
NM_001129995.1 Intron NP_001123467.1
NM_024076.2 Intron NP_076981.2
XM_011527296.1 Intron XP_011525598.1
XM_011527297.2 Intron XP_011525599.1
XM_011527298.2 Intron XP_011525600.1
XM_017027283.1 Intron XP_016882772.1
XM_017027284.1 Intron XP_016882773.1

View Full Product Details