Product Details

SNP ID
rs2117752
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:1075099 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAAGGGACAGGGCCACCGGGACAA[C/T]TCCCCGTGAGTGCGTAATTTTATGA
Phenotype
MIM: 182455
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SSTR5 PubMed Links

Gene Details

Gene
SSTR5
Gene Name
somatostatin receptor 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001053.3 Intron NP_001044.1
NM_001172560.1 Intron NP_001166031.1
Gene
SSTR5-AS1
Gene Name
SSTR5 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details