Product Details

SNP ID
rs11860141
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:67943638 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGGTGGGGGCCAAGGAAGGAGTG[C/G]TGGGGCTTGGCCCAGAGTCTGGTGT
Phenotype
MIM: 606967 MIM: 176847 MIM: 604119
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LCAT PubMed Links

Gene Details

Gene
LCAT
Gene Name
lecithin-cholesterol acyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000229.1 4480 Intron NP_000220.1
Gene
PSMB10
Gene Name
proteasome subunit beta 10
There are no transcripts associated with this gene.

Gene
SLC12A4
Gene Name
solute carrier family 12 member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145961.1 4480 UTR 3 NP_001139433.1
NM_001145962.1 4480 UTR 3 NP_001139434.1
NM_001145963.1 4480 UTR 3 NP_001139435.1
NM_001145964.1 4480 UTR 3 NP_001139436.1
NM_005072.4 4480 UTR 3 NP_005063.1

View Full Product Details