Product Details

SNP ID
rs4784780
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.16:57196675 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AATATAGGTAAGTATAGGTTATAGA[A/T]AAGTAAAAATTACTTCAGTACGGGG
Phenotype
MIM: 616585
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
RSPRY1 PubMed Links

Gene Details

Gene
RSPRY1
Gene Name
ring finger and SPRY domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001305163.1 Intron NP_001292092.1
NM_001305164.1 Intron NP_001292093.1
NM_001305182.1 Intron NP_001292111.1
NM_133368.2 Intron NP_588609.1
XM_005256220.1 Intron XP_005256277.1
XM_011523427.1 Intron XP_011521729.1
XM_011523428.1 Intron XP_011521730.1
XM_011523430.1 Intron XP_011521732.1
XM_017023844.1 Intron XP_016879333.1

View Full Product Details