Product Details

SNP ID
rs17354693
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.16:71629801 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CAGAGGCCTGTAGGTGGAACTGGCT[A/G]TGAGCTGCCAGCTGTCAGAGGAGGG
Phenotype
MIM: 614094
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MARVELD3 PubMed Links

Gene Details

Gene
MARVELD3
Gene Name
MARVEL domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017967.3 Intron NP_001017967.2
NM_001271329.1 Intron NP_001258258.1
NM_052858.5 Intron NP_443090.4
XM_011523449.2 Intron XP_011521751.1

View Full Product Details