Product Details

SNP ID
rs12596205
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:58525510 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAATTTGTGAGGTTATTTAACAAA[C/T]ACACAGATGCTCCAAAGGTGGTTGT
Phenotype
MIM: 604917 MIM: 616424
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CNOT1 PubMed Links

Gene Details

Gene
CNOT1
Gene Name
CCR4-NOT transcription complex subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001265612.1 Intron NP_001252541.1
NM_016284.4 Intron NP_057368.3
NM_206999.2 Intron NP_996882.1
Gene
SETD6
Gene Name
SET domain containing 6
There are no transcripts associated with this gene.

View Full Product Details