Product Details

SNP ID
rs2250691
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:83972196 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATGGTGAGTTTCCCTTCCAGGCC[A/G]ACGGCCGCCCCACTCCTTCTGTCCT
Phenotype
MIM: 607975
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NECAB2 PubMed Links

Gene Details

Gene
NECAB2
Gene Name
N-terminal EF-hand calcium binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019065.2 Intron NP_061938.2
XM_011523135.1 Intron XP_011521437.1
XM_017023317.1 Intron XP_016878806.1
Gene
OSGIN1
Gene Name
oxidative stress induced growth inhibitor 1
There are no transcripts associated with this gene.

View Full Product Details