Product Details
- SNP ID
-
rs8050651
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:71847541 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- ATAAATAATTGTTCTTTTTCATCTA[C/G]TGCTGCTTTTTTTCATTTAAATAAC
- Phenotype
-
MIM: 614301
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ATXN1L
PubMed Links
Gene Details
- Gene
- ATXN1L
- Gene Name
- ataxin 1 like
View Full Product Details