Product Details

SNP ID
rs13051884
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:29026015 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATACCATAAATACTGGGGAAGTTCA[C/G]TGGAGAGGTTTATTTAGAGTAGAAT
Phenotype
MIM: 604735
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
RWDD2B PubMed Links

Gene Details

Gene
RWDD2B
Gene Name
RWD domain containing 2B
There are no transcripts associated with this gene.

Gene
USP16
Gene Name
ubiquitin specific peptidase 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001992.1 Intron NP_001001992.1
NM_001032410.1 Intron NP_001027582.1
NM_006447.2 Intron NP_006438.1
XM_017028257.1 Intron XP_016883746.1
XM_017028258.1 Intron XP_016883747.1
XM_017028259.1 Intron XP_016883748.1
XM_017028260.1 Intron XP_016883749.1
XM_017028261.1 Intron XP_016883750.1

View Full Product Details