Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001007246.2 | 15028 | Intron | NP_001007247.1 | ||
NM_018963.4 | 15028 | Intron | NP_061836.2 | ||
NM_033656.3 | 15028 | UTR 3 | NP_387505.1 | ||
XM_011529611.1 | 15028 | Intron | XP_011527913.1 | ||
XM_011529612.2 | 15028 | Intron | XP_011527914.1 | ||
XM_011529613.1 | 15028 | Intron | XP_011527915.1 | ||
XM_017028373.1 | 15028 | Intron | XP_016883862.1 | ||
XM_017028374.1 | 15028 | Intron | XP_016883863.1 | ||
XM_017028375.1 | 15028 | Intron | XP_016883864.1 |