Product Details

SNP ID
rs34777077
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:1975793 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAACTTGAACCTCAGAGCACCTTGC[C/T]CTGGGTGGATGGAAGAATCCGGAAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KBTBD11 PubMed Links

Gene Details

Gene
KBTBD11
Gene Name
kelch repeat and BTB domain containing 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014867.2 Intron NP_055682.1
XM_011534771.2 Intron XP_011533073.1
XM_011534772.2 Intron XP_011533074.1
XM_017014114.1 Intron XP_016869603.1
XM_017014115.1 Intron XP_016869604.1
XM_017014116.1 Intron XP_016869605.1
XM_017014117.1 Intron XP_016869606.1
Gene
KBTBD11-OT1
Gene Name
KBTBD11 overlapping transcript 1
There are no transcripts associated with this gene.

Gene
LOC101928058
Gene Name
uncharacterized LOC101928058
There are no transcripts associated with this gene.

View Full Product Details