Product Details

SNP ID
rs1059220
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:6642664 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGACCTAACAAATATTTGAGGTCA[A/G]AACCCTACCATGTTAAAACAAACAA
Phenotype
MIM: 616905
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MRFAP1 PubMed Links

Gene Details

Gene
MRFAP1
Gene Name
Morf4 family associated protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001272053.1 2103 UTR 3 NP_001258982.1
NM_001272054.1 2103 UTR 3 NP_001258983.1
NM_033296.2 2103 UTR 3 NP_150638.1

View Full Product Details