Product Details

SNP ID
rs6446491
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:6326627 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GATCTTGCTGAAGGCATCACACATG[C/G]TCTCATCCCCAAAGCGGCCACGAAG
Phenotype
MIM: 605997
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PPP2R2C PubMed Links

Gene Details

Gene
PPP2R2C
Gene Name
protein phosphatase 2 regulatory subunit Bgamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001206994.1 Intron NP_001193923.1
NM_001206995.1 Intron NP_001193924.1
NM_001206996.1 Intron NP_001193925.1
NM_020416.3 Intron NP_065149.2
NM_181876.2 Intron NP_870991.1
XM_005247979.4 Intron XP_005248036.1
XM_011513495.1 Intron XP_011511797.1
XM_011513496.1 Intron XP_011511798.1

View Full Product Details