Product Details

SNP ID
rs11879982
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:33197813 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCAGCGGGACCCCTGCTGTGGTC[A/G]AAGGGGAGGCCCAGGGTGCACAGAA
Phenotype
MIM: 603159
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LRP3 PubMed Links

Gene Details

Gene
LRP3
Gene Name
LDL receptor related protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002333.3 Intron NP_002324.2
XM_005258945.1 Intron XP_005259002.1

View Full Product Details