Product Details

SNP ID
rs12975090
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.19:33197234 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
ATTTTCTTGAAAGGAGGGGACATGC[C/T]ACGCCATGCAGGGCCACCTGGGAAT
Phenotype
MIM: 603159
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LRP3 PubMed Links

Gene Details

Gene
LRP3
Gene Name
LDL receptor related protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002333.3 Intron NP_002324.2
XM_005258945.1 Intron XP_005259002.1

View Full Product Details