Product Details

SNP ID
rs2114358
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:128008933 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTAGATGTTTAAGCTCTTGCAGTAG[A/G]TTTTTGCAAGCTAGTGAACGCTGCC
Phenotype
MIM: 165140
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR1206 PubMed Links

Gene Details

Gene
MIR1206
Gene Name
microRNA 1206
There are no transcripts associated with this gene.

Gene
PVT1
Gene Name
Pvt1 oncogene (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details