Product Details
- SNP ID
-
rs1048166
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:41899842 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- GGAAAAACCAGTAAGAAAAGCTAAT[C/T]TTCAGGCTTCAATGTGATTTGACAT
- Phenotype
-
MIM: 605892
MIM: 605916
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
EHD4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs74573199] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- EHD4
- Gene Name
- EH domain containing 4
- Gene
- LOC105370792
- Gene Name
- uncharacterized LOC105370792
There are no transcripts associated with this gene.
- Gene
- SPTBN5
- Gene Name
- spectrin beta, non-erythrocytic 5
There are no transcripts associated with this gene.
View Full Product Details