Product Details

SNP ID
rs12466268
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:202646848 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTATGATTTTTTTTTACCTTGTTTA[A/C]GTGATAACAATAGTTTTATCCCCAG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FAM117B PubMed Links

Gene Details

Gene
FAM117B
Gene Name
family with sequence similarity 117 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173511.3 Intron NP_775782.2

View Full Product Details