Product Details

SNP ID
rs2241585
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:176529515 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCAGAGGGGTGGGAGGTGGGGCAGG[G/A]TACTCACAGCGTCTGCTCCGACTGA
Phenotype
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
RNF44 PubMed Links

Gene Details

Gene
RNF44
Gene Name
ring finger protein 44
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014901.4 Intron NP_055716.1
XM_005265840.2 Intron XP_005265897.1
XM_005265841.4 Intron XP_005265898.1
XM_005265842.4 Intron XP_005265899.1
XM_005265843.2 Intron XP_005265900.1
XM_005265844.3 Intron XP_005265901.1
XM_005265845.4 Intron XP_005265902.1
XM_006714831.2 Intron XP_006714894.1
XM_006714832.2 Intron XP_006714895.1
XM_011534466.2 Intron XP_011532768.1
XM_011534467.2 Intron XP_011532769.1

View Full Product Details